Imprimer
Chemical preservative vials, Vialservatives™
Fournisseur: EP SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
SVCN-5 SVCH-5-1 SVCN-1 SVCN-2-1 SVCS-1-1 SVCS-1-3 SVCN-2
EPSCSVCN-5EA 220 EUR
EPSCSVCN-5 EPSCSVCH-5-1 EPSCSVCN-1 EPSCSVCN-2-1 EPSCSVCS-1-1 EPSCSVCS-1-3 EPSCSVCN-2
Chemical preservative vials, Vialservatives™
Fioles
These Vialservatives™ make proper preservation at the point of sample collection convenient and safer than handling glass ampoules or larger volume chemical bottles.
  • Allows addition of preservative immediately after sample collection
  • Threaded vial containing a premeasured aliquot of preservative chemical solution
  • Individually labeled with contents and colour coded labels to minimize error in the field
Packaged in compliance with the shipping requirements of 49 CFR Part 173.4 DOT Small Quantities Classification; manufactured under an ISO certified quality management system.