Imprimer
VOA vials, economy certified
Fournisseur: EP SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
GVB-100C
EPSCGVB-100CEA 290 EUR
EPSCGVB-100C
VOA vials, economy certified
Fioles
Borosilicate glass, clear. Septa vials with all the essentials and none of the frills. The VOA vials are processed and certified to meet EPA Performance based specifications for volatile organic analysis. Boxed with a protective chip board divider to prevent breakage during transport. Vial shoulder slope optimized to prevent air bubble entrapment. Comes with open-top white polypropylene cap PTFE-lined silicone septum which is permanently bonded to the cap for a secure seal. Certificate of analysis included.
Processed to meet or exceed EPA performance based specifications for volatile organic analysis.
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable Low Manufacturing Impact
Certification environnementale NAR Certificate of Retirement