Imprimer
Water sampling bottles, glass, wide neck, amber
Fournisseur: EP SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
341-0120 341-0250 341-0500 341-0950 341-2500 341-1250 141-0060 141-0120 141-0250 141-2500 141-0500 141-0950 341-0060 241-0060 241-0120 241-0250 241-0500 241-0950
EPSC341-0120EA 112 EUR
EPSC341-0120 EPSC341-0250 EPSC341-0500 EPSC341-0950 EPSC341-2500 EPSC341-1250 EPSC141-0060 EPSC141-0120 EPSC141-0250 EPSC141-2500 EPSC141-0500 EPSC141-0950 EPSC341-0060 EPSC241-0060 EPSC241-0120 EPSC241-0250 EPSC241-0500 EPSC241-0950
Water sampling bottles, glass, wide neck, amber
Flacons Flacons d'échantillonnage de l'eau
Type III soda-lime glass, amber, with screw cap
  • With PTFE-lined white PP closed top cap (assembled on bottles)
  • Certified to meet or exceed EPA Performance Based Specifications for semi-volatile organics, pesticides, PCBs and metals analyses
  • Each jar individually barcoded with lot number and unique container number
These bottles are designed for collecting soil, sludge, waste and water samples for a variety of organic and inorganic target analytes where light protection is required. These bottles are processed the same way as the certified jars butEn savoir plus