Imprimer
Jars, sampling, wide mouth, with screw cap
Fournisseur: EP SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
120-0125 321-2000 320-0125 320-0250 320-0500 321-0125 321-0250 321-0500 321-1000 121-4000 121-0250 120-0060 121-1000 120-0250 120-0500 120-2000 121-0125 121-0500 221-2000 220-0250 220-0500 220-2000 221-0125 221-0250 220-0060 221-1000 221-4000 221-0500 220-0125 320-0060 320-2000 321-4000 V320-0125 V220-0125 V220-0250 V221-0125 V241-0950 V320-0250 V320-0500 V321-0125 V321-0250 V321-1000 V341-0950 V221-1000
EPSC120-0125EA 186 EUR
EPSC120-0125 EPSC321-2000 EPSC320-0125 EPSC320-0250 EPSC320-0500 EPSC321-0125 EPSC321-0250 EPSC321-0500 EPSC321-1000 EPSC121-4000 EPSC121-0250 EPSC120-0060 EPSC121-1000 EPSC120-0250 EPSC120-0500 EPSC120-2000 EPSC121-0125 EPSC121-0500 EPSC221-2000 EPSC220-0250 EPSC220-0500 EPSC220-2000 EPSC221-0125 EPSC221-0250 EPSC220-0060 EPSC221-1000 EPSC221-4000 EPSC221-0500 EPSC220-0125 EPSC320-0060 EPSC320-2000 EPSC321-4000 EPSCV320-0125 EPSCV220-0125 EPSCV220-0250 EPSCV221-0125 EPSCV241-0950 EPSCV320-0250 EPSCV320-0500 EPSCV321-0125 EPSCV321-0250 EPSCV321-1000 EPSCV341-0950 EPSCV221-1000
Jars, sampling, wide mouth, with screw cap
Pots Pots à usage général
Jars, clear or amber Type III soda-lime glass, with PTFE-lined white polypropylene closed-top cap, designed for collecting soil, sludge, waste and water samples for a variety of organic and inorganic target analyses.
  • Certified to meet EPA performance-based specifications for semi-volatile organics, pesticides, PCBs and metals analyses
  • Processed the same way as the certified jars but without certification documentation
  • Unprocessed and ready for your own cleaning procedure
  • Closures assembled on bottles
  • Available as short or tall profile
Informations deEn savoir plus
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact Low Manufacturing Impact
Certification environnementale NAR Certificate of Retirement NAR Certificate of Retirement NAR Certificate of Retirement