Imprimer
Water sampling bottles, HDPE, wide-mouth
Fournisseur: EP SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
313-1000 312-0120 312-0950 313-0125 313-0250 313-0500 113-0250BPC 113-0500BPC 113-1000BPC 112-2000 113-0125BPC 112-4000 312-4000BPC 112-0500 112-0120 112-0250BPC 112-0500BPC 112-0950 112-0950BPC 112-2000BPC 112-0250 112-4000BPC 312-0250BPC 312-0500 312-0500BPC 312-0950BPC 312-2000 312-2000BPC 312-4000 312-0250 312-0120BPC
EPSC313-1000EA 107 EUR
EPSC313-1000 EPSC312-0120 EPSC312-0950 EPSC313-0125 EPSC313-0250 EPSC313-0500 EPSC113-0250BPC EPSC113-0500BPC EPSC113-1000BPC EPSC112-2000 EPSC113-0125BPC EPSC112-4000 EPSC312-4000BPC EPSC112-0500 EPSC112-0120 EPSC112-0250BPC EPSC112-0500BPC EPSC112-0950 EPSC112-0950BPC EPSC112-2000BPC EPSC112-0250 EPSC112-4000BPC EPSC312-0250BPC EPSC312-0500 EPSC312-0500BPC EPSC312-0950BPC EPSC312-2000 EPSC312-2000BPC EPSC312-4000 EPSC312-0250 EPSC312-0120BPC
Water sampling bottles, HDPE, wide-mouth
Flacons Flacons d'échantillonnage de l'eau
HDPE containers, white, with LDPE-lined polypropylene cap, narrow/wide neck.
  • Narrow mouth for easy handling and precision pouring
  • Wide mouth for easy filling and sample retrieval
  • Certified to meet or exceed EPA performance-based standards for metals, cyanide and fluoride
  • Each container individually barcoded with lot number and unique container number
These containers are recommended for collecting water, leachate, and other liquid, semi-liquid, or viscous samples to be analyzed for inorganic trace analytes. Certified to meet EPA performance basedEn savoir plus