Imprimer
Cellular senescence live cell analysis assay
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
ENZKIT1300010
ENZOENZKIT1300010EA 445 EUR
ENZOENZKIT1300010
Cellular senescence live cell analysis assay
Tests Tests cellulaires
Simple assay to quantify senescence-associated β-galactosidase activity, in live cells, using flow cytometry or fluorescent microscopy.
For the study of cellular senescence in live cells.
Assay kit that uses a fluorogenic substrate to measure SA-β-gal activity in live cells.
Informations de commande: Contains Cell pretreatment solution (1000x) and SA-?-gal Substrate (200x).