Imprimer
Amp'd® HSP70 high sensitivity ELISA Kit
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
ENZKIT1010001
ENZOENZKIT1010001EA 882 EUR
ENZOENZKIT1010001
Amp'd® HSP70 high sensitivity ELISA Kit
Tests Tests ELISA
Ultra-sensitive (7 pg/ml)Amp'd® HSP70 high sensitivity ELISA kit enabling the ability to use less sample and detect both baseline and upregulated levels of human, mouse and rat Hsp70 (Hsp72), a major chaperone, cancer biomarker, and key cell stress regulator.
For the quantitative determination of normal and upregulated (stressed) levels of HSP70 in serum and plasma of human, mouse, and rat origin.
The Amp'd® HSP70 high sensitivity ELISA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4,5 hours.
Informations de commande: Contains Microtiter Plate, Assay Buffer 28, Standard, Wash Buffer Concentrate, Antibody, Conjugate, and Amp'd (Signal Amplification) Substrate, Amplifier, Substrate Diluent, Amplifier Diluent, and Stop Solution.