Imprimer
DNA methylation kit, express
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
ENZ450010050 ENZ450010200
ENZOENZ450010050EA 362 EUR
ENZOENZ450010050 ENZOENZ450010200
DNA methylation kit, express
Tests Tests à l'acide nucléique
Express DNA Methylation Kit can be used for rapid and reliable bisulfite treatment and conversion of DNA for methylation analysis.
  • High yield and ready to use
  • Easy to follow instructions
Desulphonation and clean-up of the converted DNA is performed using a unique low-elution spin column. High yield, converted DNA is ideal for PCR, array, bisulfite and next generation sequencing.
Informations de livraison: Kit contains Express conversion reagent, Methylation binding buffer, Methylation wash buffer concentrate, Methylation elution buffer , IC column, IC collection tube and L-Desulphonation reagent.