Imprimer
BIO-PROBE® Lambda probe
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
ENZ40840
ENZOENZ40840EA 420 EUR
ENZOENZ40840
BIO-PROBE® Lambda probe
Réactifs pour acides nucléiques Primers and Probes
Negative control probe for in situ hybridization.
The Lambda BIO-PROBE® labeled probe is prepared by nick translation of the lambda bacteriophage strain c1857ind1 Sam7. Fragment size range: 100-1000 base pairs (as estimated by agarose gel electrophoresis).