Imprimer
In-situ hybridisation buffer for HPV probes
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
ENZ339050050
ENZOENZ339050050EA 234 EUR
ENZOENZ339050050
In-situ hybridisation buffer for HPV probes
Tampons Tampons d'électrophorèse Tampons d'hybridation
In situ hybridisation buffer for HPV probes (ready-to use) provides a high quality hybridisation medium for In situ hybridisation analyses using either DNA or oligonucleotide probes labeled with biotin-, or fluorescein-modified nucleotides.
  • Optimised buffer to use with HPV collection of probes