Imprimer
Mycalolide B 98
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
BMLT1230020
ENZOBMLT1230020EA 394 EUR
ENZOBMLT1230020
Mycalolide B 98
Mycalolide B
Depolymerizes actin
Mycalolide B is a novel marine natural product which binds to actin in a 1:1 molar ratio (Kd=13-20 nM). It selectively and completely depolymerizes F-actin to G-actin via a mechanism distinct from that of the cytochalasins. Unlike cytochalasins, it inhibits actin-activated Mg2+-ATPase from rabbit skeletal muscle.
Isolated from the marine sponge Mycale sp.
Numéro CAS: 122752-21-0