Imprimer

Veratridine 98 TLC_ASSAY_METHOD

Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Attention

BMLNA1250010
ENZOBMLNA1250010EA 159 EUR
ENZOBMLNA1250010
Veratridine 98 TLC_ASSAY_METHOD
Veratridine

Na⁺ channel activator.


Does not affect Na⁺⁻Ca²⁺ exchange. Veratridine acts at neurotoxin receptor site 2 and preferentially binds to activated Na⁺ channels causing persistent activation. It prevents inactivation and shifts activation to more negative potentials. Induces cell death in bovine chromaffin cells.


Soluble in DMSO (25 mg/ml) or 100% ethanol (25 mg/ml).

Formule: C₃₆H₅₁NO₁₁
Poids moléculaire: 673.8 g/mol
Température de stockage: Congélateur
Numéro CAS: 71-62-5

Commander maintenant

Résultats des tests de spécification

Purity ≥98% (TLC)
Appearance White to yellow solid.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR