Imprimer

Anti-MRP4 Rat Monoclonal Antibody

Fournisseur: ENZO LIFE SCIENCES
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

ALX801039C100 ALX801039C200
ENZOALX801039C100EA 438 EUR
ENZOALX801039C100 ENZOALX801039C200
Anti-MRP4 Rat Monoclonal Antibody
Anticorps
Host: Rat, Isotype: IgG2a

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes human and mouse MRP4.

Type: Primary
Antigen: MRP4
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR