Imprimer

(2S,4R)-4-Methylglutamic acid 97

Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Synonyme(s): SYM 2081, (2S,4R)-2-Amino-4-methylpentanedioic acid

ALX550186M005
ENZOALX550186M005EA 127 EUR
ENZOALX550186M005
(2S,4R)-4-Methylglutamic acid 97
(2S,4R)-4-Methylglutamic acid

Potent and highly selective kainate receptor agonist, with an IC50 for inhibition of [3H]-kainate binding of 35 nM and almost 3,000- and 200-fold selectivity for kainate receptors over AMPA and NMDA receptors respectively. Also selectively inhibits the cloned excitatory amino acid transporter EAAT2 at higher concentrations.


  • Kainate receptor agonist


White powder.


Soluble in dilute aqueous acid.

Formule: C₆H₁₁NO₄
Poids moléculaire: 161.2 g/mol
Température de stockage: Congélateur
Numéro MDL: MFCD00937765
Numéro CAS: 31137-74-3

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR