Imprimer

KN-93 (water soluble) ≥98% (par 1H-RMN)

Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Synonyme(s): (E)-N-(2-(((3-(4-Chlorophenyl)allyl)(methyl)amino)methyl)phenyl)-N-(2-hydroxyethyl)-4-methoxybenzenesulphonamide phosphate, (E)-KN-93 phosphate

ALX430127M001 ALX430127M005
ENZOALX430127M001EA 162 EUR
ENZOALX430127M001 ENZOALX430127M005
KN-93 (water soluble) ≥98% (par 1H-RMN)
KN-93 phosphate

CaM kinase II inhibitor


Selective inhibitor of rat brain Ca²⁺/calmodulin-dependent protein kinase II. Inhibits glucose-induced and forskolin-stimulated insulin release from isolated pancreatic islets of Langerhans. Has no significant effect on protein kinase A (PKA) activity. Induces G1 cell cycle arrest and apoptosis in NIH 3T3 cells.

Formule: C₂₆H₂₉ClN₂O₄S·H₃PO₄
Poids moléculaire: 501.0·98.0 g/mol
Numéro MDL: MFCD03788201
Numéro CAS: 1188890-41-6 | 139298-40-1 (free base)

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR