Imprimer

Polymyxine B, sulfate

Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Attention

ALX380040G001 ALX380040G005
ENZOALX380040G001EA 120 EUR
ENZOALX380040G001 ENZOALX380040G005
Polymyxine B, sulfate
Polymyxine B, sulfate

PKC inhibitor


Antibiotic. Inhibitor of protein kinase C (PKC). Breaks the bacterial membrane by incorporating into the phospholipid of the outer membrane and activating phospholipase.


Isolated from Bacillus polymyxa.

Formule: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
Poids moléculaire: 1385.61 g/mol
Température de stockage: Température ambiante
Numéro MDL: MFCD00131991
Numéro CAS: 1405-20-5
EINECS: 215-774-7

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR