Imprimer

Baccatine III 95 HPLC_ASSAY_METHOD

Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Danger

ALX351005M025
ENZOALX351005M025EA 407 EUR
ENZOALX351005M025
Baccatine III 95 HPLC_ASSAY_METHOD
Baccatine III

Antitumor agent.


Starting material for the synthesis of taxol analogs. Shows antitumor properties.


Soluble in DMSO or 100% ethanol. Avoid strong oxidising agents, acids and bases.

Formule: C₃₁H₃₈O₁₁
Poids moléculaire: 586.634 g/mol
Température de stockage: Congélateur
Numéro CAS: 27548-93-2

Commander maintenant

Résultats des tests de spécification

Purity ≥95% (HPLC)
Source Semisynthetic from Taxus baccata


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR