Imprimer
Oxamflatin 95 HPLC_ASSAY_METHOD
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
ALX270379M005
ENZOALX270379M005EA 527 EUR
ENZOALX270379M005
Oxamflatin 95 HPLC_ASSAY_METHOD
Oxamflatin
HDAC inhibitor
Potent inhibitor of mammalian HDACs (histone deacetylases) (IC50=15.7nM). Acts as a ligand for the enzyme active site metal ion. Upregulates plasminogen activator inhibitor type 2 (PAI-2) expression with concomitant inhibition of urokinase (u-PA) gene and protein expression in HT-1080 and U-937 cells. Elevates the expression of extracellular matrix proteins fibronectin and gelsolin. Induces apoptosis in P-glyoprotein (Pgp) positive and Pgp negative multidrug resistant cells.
Numéro CAS: 151720-43-3