Imprimer
Ac-VEID-AMC
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
ALX260064M001 ALX260064M005
ENZOALX260064M001EA 77 EUR
ENZOALX260064M001 ENZOALX260064M005
Ac-VEID-AMC
Protéines et Peptides
Caspase-6 substrate
Fluorogenic substrate for caspase-6 (Km=30µML, Kcat/Km=168000M-1sec-1) and related cysteine proteases. Sequence is based on lamin A site of cleavage. Ex.:340 to 360 nm, Em.:440 to 460 nm.