Imprimer

Anti-CASP1 Rabbit Polyclonal Antibody

Fournisseur: ENZO LIFE SCIENCES
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

ALX210804C100
ENZOALX210804C100EA 493 EUR
ENZOALX210804C100
Anti-CASP1 Rabbit Polyclonal Antibody
Anticorps
Host: Rabbit

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse and rat caspase-1. Detects bands of ~45kDa (procaspase-1) and ~20kDa (cleavage product) by Western blot.

Type: Primary
Antigen: CASP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR