Imprimer
HSP70 ELISA Kit
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
ADIEKS700B
ENZOADIEKS700BEA 812 EUR
ENZOADIEKS700B
HSP70 ELISA Kit
Tests Tests ELISA
Widely used ELISA HSP70 kit for chaperone and stress research.
For the quantitative determination of HSP70 in cell lysates and tissue of human, mouse, and rat origin. Prod. No. ADI-EKS-715 is recommended for the detection of Hsp70 in serum and plasma samples.
The HSP70 EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in 4,5 hours. Absorbance is read at 450 nm. This kit is ideal for measuring intracellular HSP70 (HSP72). Save time, money, and precious sample with fully quantitative results and increased sensitivity compared to Western blot analysis.
Informations de commande: Contains Microtiter plate, 5X Extraction reagent, Standard, Sample diluent 2, Wash buffer concentrate, Antibody, Conjugate, TMB Substrate, Stop solution 2.