Imprimer
Blue IHC chromogen (HRP), HIGHDEF®
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
ADI9501510030
ENZOADI9501510030EA 245 EUR
ENZOADI9501510030
Blue IHC chromogen (HRP), HIGHDEF®
Tests Tests cellulaires
Blue IHC chromogen (HRP) is intended to be used in conjunction with horseradish peroxidase (HRP)-based immunostaining or in situ hybridisation systems.
  • Unique high definition blue IHC chromogen for HRP-activated development
  • Produces a sharply contrasting billiant blue colour
  • Easily be distinguished from other stains
Informations de livraison: Substrate buffer and chromogen in an empty mixing bottle.