Imprimer
Chemiluminescent acridinium esters
Marques : ENZO LIFE SCIENCES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
ADI9060001 ADI906001
ENZOADI9060001EA 503 EUR
ENZOADI9060001 ENZOADI906001
Chemiluminescent acridinium esters
Réactifs d'électrophorèse Western Blotting Reagents
Chemiluminescent acridinium esters are commonly used to label antibodies, proteins, nucleic acids, and some peptides, and are frequently used as detection reagents in immunoassays.
  • Species independent
New Product
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable