Imprimer
Film frame rings for wafers
Fournisseur: ENTEGRIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
FFP200276-4602 FFP150217-1904 FFP3003804602
ENTEFFP200276-4602EA 0 EUR
ENTEFFP200276-4602 ENTEFFP150217-1904 ENTEFFP3003804602
Film frame rings for wafers
Scellés Bagues d'étanchéité
Plastic film frame rings provide a cost-effective and light weight alternative to expensive, heavy, metal frames to handle wafers during assembly operations and can be used to store and ship finished wafers.
  • Film frames rings are available for 300, 200 and 150 mm wafers
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable