Imprimer
Pneumatic valves, CG4 SG series
Fournisseur: ENTEGRIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
CR4-2C-4N CR4-2C-4F CR4-3C-6F-3 CR4-2C-4F-3 CR4-2C-6F CR4-2CL-4F-3 CR4-2CL-6F-3 CR4-2U-4N CR4-2UL-4F-3 CR4-2UL-6F-3
ENTECR4-2C-4NEA 0 EUR
ENTECR4-2C-4N ENTECR4-2C-4F ENTECR4-3C-6F-3 ENTECR4-2C-4F-3 ENTECR4-2C-6F ENTECR4-2CL-4F-3 ENTECR4-2CL-6F-3 ENTECR4-2U-4N ENTECR4-2UL-4F-3 ENTECR4-2UL-6F-3
Pneumatic valves, CG4 SG series
Vannes Micro-vannes
CR4 pneumatic valves are specially designed to handle temperatures upto 160 °C. There is a option to add ¼" and ¹/₈" connections from Flaretek®. With no exposed metal hardware the valve is completely sealed and protected from harsh chemical environments.
  • For corrosive environments in wet clean applications
  • Transporting and protecting high-purity chemicals
  • Chemical line size in ³/₈" or smaller