Imprimer
Test sieves, 100×40 mm
Fournisseur: ENDECOTTS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
100SIW.063 100SIW.150 100SIW.425 100SIW.355 100SIW.300 100SIW.032 100SIW.036 100SIW.038 100SIW.125 100SIW.250 100SIW.045 100SIW.280 100SIW.100 100SIW.160 100SIW.040 100SIW.200 100SIW.400 100SIW1.00
ENDE100SIW.063EA 243 EUR
ENDE100SIW.063 ENDE100SIW.150 ENDE100SIW.425 ENDE100SIW.355 ENDE100SIW.300 ENDE100SIW.032 ENDE100SIW.036 ENDE100SIW.038 ENDE100SIW.125 ENDE100SIW.250 ENDE100SIW.045 ENDE100SIW.280 ENDE100SIW.100 ENDE100SIW.160 ENDE100SIW.040 ENDE100SIW.200 ENDE100SIW.400 ENDE100SIW1.00
Test sieves, 100×40 mm
Tamis Tamis analytiques
Stainless steel woven wire mesh test sieves are ideal for all types of laboratory sampling and particle size analysis.
For more size options please contact your local Avantor office.
Certifications: ISO3310-1:2000, BS410-1:2000
Informations de livraison: Each sieve is supplied together with a certificate of examination and carries the BS 410 kite mark.