Imprimer
Pipette tracker, LightOne™ Illuminator qRED, Embitec®
Fournisseur: EMBITEC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
LI-1300
EMBILI-1300EA 606 EUR
EMBILI-1300
Pipette tracker, LightOne™ Illuminator qRED, Embitec®
Transilluminateurs Transilluminateurs LED
Compact benchtop illuminator helps to visually identify a specific sample well, complete lanes or rows, to aid in loading 96-well plates.
  • Red LEDs minimize quenching of fluorescent dyes, with three distinct intensities
  • Auto and manual/hands-free advance modes
  • Dedicated LED for each sample well to provide accurate guidance
  • User defined time intervals for advancing to the next sample(s)
  • Illuminate single well or complete lane or complete row
  • Material: White polycarbonate
The LightOne™ Illuminator qRED is a stand-alone bench top solution for trackingEn savoir plus