Imprimer
Insoles, low arch support, 8711L, JALAS®
Fournisseur: EJENDALS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
8711L-34 8711L-35 8711L-36 8711L-37 8711L-38 8711L-39 8711L-40 8711L-41 8711L-42 8711L-43 8711L-44 8711L-45 8711L-46 8711L-47 8711L-48 8711L-49 8711L-50
EJEN8711L-34EA 38.6 EUR
EJEN8711L-34 EJEN8711L-35 EJEN8711L-36 EJEN8711L-37 EJEN8711L-38 EJEN8711L-39 EJEN8711L-40 EJEN8711L-41 EJEN8711L-42 EJEN8711L-43 EJEN8711L-44 EJEN8711L-45 EJEN8711L-46 EJEN8711L-47 EJEN8711L-48 EJEN8711L-49 EJEN8711L-50
Insoles, low arch support, 8711L, JALAS®
Semelles de chaussure
Insoles for low arch support made from textile, soft EVA polyester based electro conductive thread and TPU control bar.
  • Anatomically designed and anti-static properties
  • Double shock absorption zones in PORON® XRD®
  • Perfect fit with extra comfortable and breathable
These ergonomically shaped insoles are suitable for use in all JALAS® shoes.
Colour: Orange/black
Commander maintenant
SPÉCIFICATIONS
Environmentally Preferable