Imprimer
Highlight Jalas® 3498 Safety Ankle Boots
Fournisseur: EJENDALS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
3498-46 3498-36 3498-37 3498-38 3498-39 3498-40 3498-41 3498-42 3498-43 3498-44 3498-45 3498-47
EJEN3498-46EA 144.7 EUR
EJEN3498-46 EJEN3498-36 EJEN3498-37 EJEN3498-38 EJEN3498-39 EJEN3498-40 EJEN3498-41 EJEN3498-42 EJEN3498-43 EJEN3498-44 EJEN3498-45 EJEN3498-47
Highlight Jalas® 3498 Safety Ankle Boots
Chaussures et bottes
Anti-static safety ankle boot made from water repellent PU-coated leather/textile combination and a oil and slip resistant PU out sole.
  • Polyester/polyamide lining
  • Shock absorption zone
  • Puncture resistant plasma-treated composite (PTC) textile midsole
  • Protective aluminium toecap (200 J)
For use in harsh environments, environments with risk for outsole penetration, all-year outdoor use.
Colour: Black
Certifications: EN ISO 20345:2011; S3, SRC