Imprimer
ESD safety shoes, lace-up, Zenit, JALAS® 3030
Fournisseur: EJENDALS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
3030-36 3030-37 3030-38 3030-39 3030-40 3030-41 3030-42 3030-43 3030-44 3030-45 3030-46 3030-47
EJEN3030-36EA 148.68 EUR
EJEN3030-36 EJEN3030-37 EJEN3030-38 EJEN3030-39 EJEN3030-40 EJEN3030-41 EJEN3030-42 EJEN3030-43 EJEN3030-44 EJEN3030-45 EJEN3030-46 EJEN3030-47
ESD safety shoes, lace-up, Zenit, JALAS® 3030
Chaussures et bottes
Anti-static ESD safety shoes made from PU-coated and split leather with EVA, polyester based conductive thread insole and merino wool lining.
  • Oil resistant outsole
  • Double shock absorption zones in Poron® XRD
  • Padded shaft edge made from memory foam with ventilating insole
  • Puncture resistant midsole made from plasma-treated composite (PTC) textile
  • Toe protection cap made from aluminium (200 J)
Mainly used for outdoors and environments with risk for outsole penetration.
Colour: Black/grey/red
Certifications: EN 61340-5-1 (ESD); EN ISO 20345:2011; S2 SRC