Imprimer
Floor Stand for 30 L Bio-bin® Boxes
Fournisseur: ECONIX LTD
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
EXBBS30.0 EXBBSW30.0
ECONEXBBS30.0EA 120 EUR
ECONEXBBS30.0 ECONEXBBSW30.0
Floor Stand for 30 L Bio-bin® Boxes
Boîtes Boîtes à déchets
This stainless steel stand is perfect for lifting the 30 Litre Bio-bin® off the floor and holding it firmly in an upright position.
  • Bio-bin® can be easily accessed, sealed down and removed for disposal
  • Four adjustable feet: Stand will not damage the floor and can accommodate uneven surfaces
  • Available with or without wheels