Imprimer
VWR®, EDTA buffer solution pH 8,0 (0,5 mol/L) for biotechnology, sterile
Marques : VWR Chemicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
E522-100MLEA 105 EUR
E522-100ML
VWR®, EDTA buffer solution pH 8,0 (0,5 mol/L) for biotechnology, sterile
Tampons Tampons de clonage