Imprimer
Safety cabinets for pressurised gas cylinders, type G30, ECO line plus
Fournisseur: DUPERTHAL
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
73-200960-011 73-201260-011 73-200660-011
DUEH73-200960-011EA 2830 EUR
DUEH73-200960-011 DUEH73-201260-011 DUEH73-200660-011
Safety cabinets for pressurised gas cylinders, type G30, ECO line plus
Mobilier Armoires de stockage
Safety cabinets for pressurised gas cylinders-inside storage is made up of sheet steel, powder-coated.
  • Classical door technology
  • Access control with cylinder lock
  • Enhanced ventilation in every cabinet level
  • Adjustable feet for level equalisation
Certifications: Conforms with DIN EN 14470-2, EK5/AK4 09-10, DIN EN 14727, CE certified, TRGS 510 chapter 10, type G30.