Imprimer
Multi-gas detector, Accuro®
Fournisseur: Dräger
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
8318392
DRAE8318392EA 787 EUR
DRAE8318392
Multi-gas detector, Accuro®
Systèmes de détection de gaz
Specifically designed for fast measurements with one hand, this gas detector pump allows to use tested tubes under extreme conditions.
Informations de commande: Includes hand bellows pump and spares kit in standard leather carrying case.