Imprimer
Microtube membranes, breathable, Breathe-EASIER
Fournisseur: DIVERSIFIED BIOTECH
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
BMTM-2000
DIVEBMTM-2000EA 152 EUR
DIVEBMTM-2000
Microtube membranes, breathable, Breathe-EASIER
Scellés Films de scellage
Breathe-EASIER micropermeable membranes are ideal for cell growth, concentrating samples, and gas exchange. Formatted with an easy to use finger tab, membranes are precut to fit a variety of tube sizes. Membranes are gamma irradiated to ensure sterility and are free of cytotoxins.
  • Microtube cap closures will not bind in centrifuge rotors
  • Convenient scoring makes liner removal quick and easy
  • White non woven rayon membrane rapidly transmits water vapour for accelerated growth