Imprimer
Sealable Standup Pouches
Fournisseur: DAKLAPACK
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
481121 481125
DAKL481121EA 106 EUR
DAKL481121 DAKL481125
Sealable Standup Pouches
Sachets Sacs de stockage
These pouches are intended for use as a one time use pouch for food and even non food products. The pouch is designed with an opening bottom gusset which means the pouch will stand up on its own.
  • Available in many different sizes
  • High quality laminated foil offers high barrier properties
  • With a round bottom gusset and an opening without zip
Solids, liquids or powders can all be store in these pouches and easily sealed closed using a heat sealer.