Imprimer
Adjustable wristbands, DBI-SALA®
Fournisseur: CAPITAL SAFETY
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
1500085 1500087
CPSD1500085EA 0 EUR
CPSD1500085 CPSD1500087
Adjustable wristbands, DBI-SALA®
Dispositifs de protection antistatique Bracelets antistatiques
Adjustable wristbands are available with retractor and elastic strap with hook and loop, or with cord and elastic strap with hook and loop.
  • Adjusts to virtually any wrist size with hook and loop system
  • Replaceable steel cable retractor fits into a sewn-in pocket and allows tools to easily be tied off using an attached thumb-operated trigger
  • Elastic band helps ensure a comfortable fit
  • Attached trigger can be used to quickly work with many hand tools
The DBI-SALA® Adjustable wristband with retractor is constructed out of elastic webbing that can beEn savoir plus