Imprimer
Cleaning swabs, double-ended, CONSTIX® SC-1
Fournisseur: CONTEC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
SC-1
CONTSC-1EA 596 EUR
CONTSC-1
Cleaning swabs, double-ended, CONSTIX® SC-1
Écouvillons Écouvillons de nettoyage
SC-1 double-ended pointed cotton swabs consist of high quality, long staple-length cotton, tightly wrapped and shaped on both ends of high strength, wound paper handles.
  • Medium, pointed cotton ends
  • High-quality cotton fibers provide excellent particle entrapment and retention
  • Tightly wrapped head help maintain low levels of extractable residue
  • Heads are held to the handles using a methyl-cellulose binder
The high-quality cotton fibers provide exceptional sorbency of most solvents and solutions. The heads are very tightly wrapped and shaped using aEn savoir plus