Imprimer
Bond tack cloth
Fournisseur: CONTEC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
BS20CB-12
CONTBS20CB-12EA 651 EUR
CONTBS20CB-12
Bond tack cloth
Lingettes Chiffons nettoyants réutilisables Chiffons pour tâches délicates
These polyester cloths are ideal for the surface preparation prior to painting. Also suitable for particle removal in printed circuit board manufacturing and for eliminating solvents in some applications.
  • Custom blended tack formula for optimal adhesive texture, transfer resistance, heat resistance and anti-static performance
  • Sealed edge technology minimises particles and fibres
  • "Napped" surface easily removes dirt particles
  • Water-based tack is applied at high heat under pressure