Imprimer
Adaptor, QuickConnect™
Fournisseur: CONTEC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
2758
CONT2758EA 90.3 EUR
CONT2758
Adaptor, QuickConnect™
Serpillières
Stainless steel adaptor for use with QuickTask™ mops.
  • Autoclavable at 121 °C for 30 minutes
The QuickConnect™ adaptor allows users to connect any QuickConnect™ handle to the QuickTask™ polypropylene mop frame.