Imprimer
Pallets and sumps, PE
Fournisseur: CEMO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
10019 10017 10018 8524 10285
CEMG10019EA 0 EUR
CEMG10019 CEMG10017 CEMG10018 CEMG8524 CEMG10285
Pallets and sumps, PE
Stockage des fûts
These sumps are manufactured from high quality polyethylene (HD-LMD).
  • With general construction inspection approval no. Z-40.22-380
  • High chemical resistance
Sump pallets 240 have removable perforated plate and the forklift-accessible on all four sides is possible only in 240/4.
Sump pallet 230/2 are suitable for Euro-pallets (1200×800 mm) and plastic pallets of similar size. These are loaded pallets may be placed by a forklift directly on the tray (PE perforated plate is not required).