Imprimer
Mobile Boxes for Hazardous Materials
Fournisseur: CEMO
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
7718 7805 7719 8318 7720 7806
CEMG7718EA 289 EUR
CEMG7718 CEMG7805 CEMG7719 CEMG8318 CEMG7720 CEMG7806
Mobile Boxes for Hazardous Materials
Boîtes Boîtes à déchets
These mobile boxes with its stable design enables clean and safe storage of hazardous solid goods belonging to packing groups II or III and is also approved for transport according to ADR, RID and IMDG codes.
  • Easy to handle, with coloured lids
  • Specially shaped lid enables storage in two-layers
  • Up to eight boxes can be safely stacked on a Euro pallet
  • Beading in the container wall enables emptying with a rotary forklift