Imprimer

Peroxynitrite Na dans une solution sodium hydroxyde

Marques : Cayman Chemical
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

Attention

Synonyme(s): Oxido nitrite, Peroxynitrous acid sodium salt, Peroxynitrite, Sodium peroxynitrite

81565-1
CAYM81565-1EA 72.6 EUR
CAYM81565-1
Peroxynitrite Na dans une solution sodium hydroxyde
Peroxynitrite Na

Peroxynitrite is formed in vivo by the reaction of NO with superoxide. It is a powerful oxidizing agent that can initiate lipid peroxidation, oxidize sulfhydryls, and nitrate the aromatic residues of proteins.


  • Purity: ≥90% (balance is NO2/NO3)
  • Formulation: A solution in 0.3 M sodium hydroxide
  • λmax: 302 nm

Formule: ONO₂·Na
Poids moléculaire: 85.0 g/mol
Température de stockage: Congélateur ultra-basse température (–80°C)
Numéro CAS: 14042-01-4

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR