Imprimer

Resolvin D2 ELISA kit

Marques : Cayman Chemical
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

501120-480 501120-96 501120-96S 501120-480S
CAYM501120-480EA 2730 EUR
CAYM501120-480 CAYM501120-96 CAYM501120-96S CAYM501120-480S
Resolvin D2 ELISA kit
Tests Tests ELISA

Resolvin D2 ELISA kit is a competitive assay that can be used for quantification of RvD2.


  • Detect RvD2 in multiple sample types
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Measure RvD2 levels down to 10 pg/ml
  • Reads colorimetric at 405 to 420 nm


Resolvin D2 (RvD2) is produced physiologically from the sequential oxygenation of docosahexaenoic acid (DHA) by 15- and 5-lipoxygenases. RvD2 reduces polymorphonuclear leukocyte transendothelial migration, the earliest event in acute inflammation, by ~70% at doses as low as 10 pg in the mouse model of peritonitis. In the rat model of burn injury and sepsis, RvD2 restores defective neutrophil motility and dramatically increases survival at doses of 8 to 10 ng/rat. Resolvins also enhance bacterial clearance by phagocytosis and have been shown to synergistically enhance certain antibiotics.


Due to the number and variation of potential sample types, this assay has been validated in Cayman’s ELISA Buffer (Item No. 400060 diluted to 1X (0.1 M potassium phosphate containing 0.1% BSA, 400 mM sodium chloride, 1 mM EDTA, and 0.01% sodium azide), human plasma, and human serum.


Cdt: Kit contains: Antiserum, tracer, standard, ELISA and wash buffer concentrate, polysorbate 20, plate and cover, Ellman's reagent, tracer and antiserum dye.

Commander maintenant

SPÉCIFICATIONS

Environmentally Preferable
Primary antibody reactivity
Hôte
Sensibilité 10 pg/ml (80% B/B₀)
Portée de détection 1,6 - 1000 pg/ml
Milieu 20 - 60 pg/ml (50% B/B₀)
Temps de développement 90 - 120 min
Temps d'incubation 18 hr
Conditions de stockage –80 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR