Imprimer

PAD4 Autoantibody ELISA kit

Marques : Cayman Chemical
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

500930-96
CAYM500930-96EA 779 EUR
CAYM500930-96
PAD4 Autoantibody ELISA kit
Tests Tests ELISA

An immunometric for the measurement of anti-PAD4 autoantibodies.


  • Measure anti-PAD4 autoantibodies of any isotype (IgM, IgG, IgA)
  • For use in human plasma and serum (no need to purify samples)
  • Lower Limit of quantification is 31,3 U/ml


PAD4 is a guanidino-modifying enzyme which catalyzes the conversion of specific arginine residues to citrulline in a calcium-dependent manner. PAD4 activity is increased in rheumatoid arthritis (RA), producing citrulline-containing proteins (CCP) that induce the formation of arthritogenic autoantibodies.


In addition to anti-CCP autoantibodies, 23 to 45% of RA patients also produce autoantibodies specific for the PAD4 enzyme itself. PAD4 Autoantibody ELISA Kit is an immunometric assay which can be used to measure anti-PAD4 autoantibodies of any isotype (IgM, IgG, and IgA) in human plasma and serum without prior sample purification.


Affinity-purified anti-PAD4 antibody isolated from the plasma of a patient with RA is used as the standard. One unit is approximately equal to 1 ng of anti-PAD4 Ig protein.


Cdt: Kit contains: HRP conjugate, plate, ELISA standard, wash buffer concentrate, non-fat dry milk assay reagent, ELISA buffer concentrate, polysorbate 20, substrate and stop solution, cover sheet.

Commander maintenant


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR