Imprimer

Anti-TRAIL-R4 Rabbit Polyclonal Antibody

Fournisseur: Cayman Chemical
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

160755-1
CAYM160755-1EA 648 EUR
CAYM160755-1
Anti-TRAIL-R4 Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: TRAIL-R4
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR