Imprimer

Anti-MPO Mouse Monoclonal Antibody [clone: 5F6]

Fournisseur: Cayman Chemical
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

15640-1
CAYM15640-1EA 498 EUR
CAYM15640-1
Anti-MPO Mouse Monoclonal Antibody [clone: 5F6]
Anticorps
Type: Primary
Antigen: MPO
Clonality: Monoclonal
Clone: 5F6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR