Imprimer

Anti-CD25 PE Mouse Monoclonal Antibody [clone: 143-13]

Fournisseur: Cayman Chemical
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : sur 5

10339-1
CAYM10339-1EA 556 EUR
CAYM10339-1
Anti-CD25 PE Mouse Monoclonal Antibody [clone: 143-13]
Anticorps
Type: Primary
Antigen: CD25 PE
Clonality: Monoclonal
Clone: 143-13
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR