Imprimer
Industrial Bucket
Fournisseur: BURKLE
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
2308-0900 2308-1700 2308-1200
BURK2308-0900EA 16 EUR
BURK2308-0900 BURK2308-1700 BURK2308-1200
Industrial Bucket
Godets
LLDPE, white.
  • With litre scale, graduation 1000 ml
  • With spout
  • Suitable for use with foodstuffs
Industrial bucket for transporting, decanting and mixing. Safe emptying with wide spout. Handle made of metal.