Imprimer
Kjeldahl digestion system, KjelDigester K-446
Fournisseur: BUCHI
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : sur 5
1154461000
BUCH1154461000EA 12190 EUR
BUCH1154461000
Kjeldahl digestion system, KjelDigester K-446
Systèmes de digestion Systèmes de minéralisation
The KjelDigester provides fast and safe digestion of up to 20 samples simultaneously.
  • Fully automated
  • High throughput nitrogen and protein determination
  • Convenient automatic lift for raising and lowering the sample tube rack
  • Programming mode to save all digestion methods